missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indocyanine green, MP Biomedicals™
102.45 € - 200.34 €
Chemical Identifiers
CAS | 3599-32-4 |
---|---|
Molecular Formula | C43H47N2NaO6S2 |
Molecular Weight (g/mol) | 774.967 |
MDL Number | MFCD00013078 |
InChI Key | MOFVSTNWEDAEEK-UHFFFAOYSA-M |
Synonym | Cardiogreen, Fax green |
PubChem CID | 132274068 |
ChEBI | CHEBI:31696 |
IUPAC Name | sodium;4-[(2E)-2-[(2E,4E,6Z)-7-[1,1-dimethyl-3-(4-sulfonatobutyl)benzo[e]indol-3-ium-2-yl]hepta-2,4,6-trienylidene]-1,1-dimethylbenzo[e]indol-3-yl]butane-1-sulfonate |
SMILES | CC1(C(=[N+](C2=C1C3=CC=CC=C3C=C2)CCCCS(=O)(=O)[O-])C=CC=CC=CC=C4C(C5=C(N4CCCCS(=O)(=O)[O-])C=CC6=CC=CC=C65)(C)C)C.[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11413222
|
MP Biomedicals
0215502025 |
25 mg |
102.45 €
25mg |
Estimated Shipment: 07-10-2024
Log in to see stock. |
|||||
11423222
|
MP Biomedicals
0215502050 |
50 mg |
159.00 €
50mg |
Estimated Shipment: 15-10-2024
Log in to see stock. |
|||||
11433222
|
MP Biomedicals
0215502080 |
100 mg |
200.34 €
100mg |
Estimated Shipment: 13-11-2024
Log in to see stock. |
|||||
Description
Chemical Identifiers
3599-32-4 | |
774.967 | |
MOFVSTNWEDAEEK-UHFFFAOYSA-M | |
132274068 | |
sodium;4-[(2E)-2-[(2E,4E,6Z)-7-[1,1-dimethyl-3-(4-sulfonatobutyl)benzo[e]indol-3-ium-2-yl]hepta-2,4,6-trienylidene]-1,1-dimethylbenzo[e]indol-3-yl]butane-1-sulfonate |
C43H47N2NaO6S2 | |
MFCD00013078 | |
Cardiogreen, Fax green | |
CHEBI:31696 | |
CC1(C(=[N+](C2=C1C3=CC=CC=C3C=C2)CCCCS(=O)(=O)[O-])C=CC=CC=CC=C4C(C5=C(N4CCCCS(=O)(=O)[O-])C=CC6=CC=CC=C65)(C)C)C.[Na+] |
Specifications
3599-32-4 | |
C43H47N2NaO6S2 | |
Cardiogreen, Fax green | |
CC1(C(=[N+](C2=C1C3=CC=CC=C3C=C2)CCCCS(=O)(=O)[O-])C=CC=CC=CC=C4C(C5=C(N4CCCCS(=O)(=O)[O-])C=CC6=CC=CC=C65)(C)C)C.[Na+] | |
774.967 | |
CHEBI:31696 | |
Green | |
Indocyanine green |
∽ 90% | |
MFCD00013078 | |
MOFVSTNWEDAEEK-UHFFFAOYSA-M | |
sodium;4-[(2E)-2-[(2E,4E,6Z)-7-[1,1-dimethyl-3-(4-sulfonatobutyl)benzo[e]indol-3-ium-2-yl]hepta-2,4,6-trienylidene]-1,1-dimethylbenzo[e]indol-3-yl]butane-1-sulfonate | |
132274068 | |
774.98 | |
Crystalline Powder |
Safety and Handling
Recommended Storage : Store at Room Temperature (15 to 30°C). Desiccate.
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- INDOCYANINE GREEN
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : indocyanine green