missing translation for 'onlineSavingsMsg'
Learn More

Indocyanine green, MP Biomedicals™

Product Code. 11423222
Click to view available options
Quantity:
25 mg
50 mg
100 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 3599-32-4
Molecular Formula C43H47N2NaO6S2
Molecular Weight (g/mol) 774.967
MDL Number MFCD00013078
InChI Key MOFVSTNWEDAEEK-UHFFFAOYSA-M
Synonym Cardiogreen, Fax green
PubChem CID 132274068
ChEBI CHEBI:31696
IUPAC Name sodium;4-[(2E)-2-[(2E,4E,6Z)-7-[1,1-dimethyl-3-(4-sulfonatobutyl)benzo[e]indol-3-ium-2-yl]hepta-2,4,6-trienylidene]-1,1-dimethylbenzo[e]indol-3-yl]butane-1-sulfonate
SMILES CC1(C(=[N+](C2=C1C3=CC=CC=C3C=C2)CCCCS(=O)(=O)[O-])C=CC=CC=CC=C4C(C5=C(N4CCCCS(=O)(=O)[O-])C=CC6=CC=CC=C65)(C)C)C.[Na+]

Specifications

Assay Percent Range ∽ 90%
Formula Weight 774.98
Color Green
Quantity 50 mg
Physical Form Crystalline Powder
Chemical Name or Material Indocyanine green
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'