missing translation for 'onlineSavingsMsg'
Learn More

Brand™ Silberbrand™ Imhoff Sedimentation Cones

Manufactured from borosilicate glass and complies with DIN 12672 standards. BRAND™ Silberbrand™ Imhoff Sedimentation Cones are available with or without stopcock, graduated to 100mL or 1000mL, and include a ring mark at 1000mL. *1000mL graduated version does not include ring mark.

167.00 € - 399.00 €

Specifications

Material Borosilicate Glass
Product Type Sedimentation cone
View More Specs

Products 3
Product Code Brand Graduated Description Price Quantity & Availability  
Product Code Brand Graduated Description Price Quantity & Availability  
11824060
View Documents Promotion Details
Brand™
387462
to 1000mL Without stopcock, no ring mark This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10028840
View Documents Promotion Details
Brand™
387662
to 100mL With stopcock, ring mark at 1000mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10664141
View Documents Promotion Details
Brand™
387362
to 100mL Without stopcock, ring mark at 1000mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Manufactured from borosilicate glass.
  • Complies with DIN 12672 standards.
  • With or without stopcock
  • Graduated to 100mL or 1000mL
  • Ring mark at 1000mL
  • *1000mL graduated version does not include ring mark.
Specifications

Specifications

Borosilicate Glass
Sedimentation cone
Videos
SDS
Documents

Documents

Product Certifications