missing translation for 'onlineSavingsMsg'
Learn More

Brand™ Silberbrand™ Imhoff Sedimentation Cones

Manufactured from borosilicate glass and complies with DIN 12672 standards. BRAND™ Silberbrand™ Imhoff Sedimentation Cones are available with or without stopcock, graduated to 100mL or 1000mL, and include a ring mark at 1000mL. *1000mL graduated version does not include ring mark.

Brand:  Brand™ 387362

 View more versions of this product

Product Code. 10664141

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Manufactured from borosilicate glass.
  • Complies with DIN 12672 standards.
  • With or without stopcock
  • Graduated to 100mL or 1000mL
  • Ring mark at 1000mL
  • *1000mL graduated version does not include ring mark.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Borosilicate Glass
Without stopcock, ring mark at 1000mL
to 100mL
Sedimentation cone
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers