Learn More
Sampling Systems™ Ice Borer
Constructed with sharp cutting teeth, which allows the device to be suitable for use on frozen products such as meats. Sampling Systems™ Ice Borer is a robust and efficient sampler that is ideal for sampling ice and waxy products.
Brand:  Sampling Systems™ 6005A-500
 View more versions of this product
Product Code. 15889371
  • 143.00 € / Each
Estimated Shipment: 04-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Suitable for ice, wax and other similar materials
  • 316 Stainless Steel
  • Rotate the sampler as it is pushed into the product
  • The sharp edge will cut into the product and the sample will be deposited in the tube
  • Push the discharge rod down the Ice Borer to remove the sample
Specifications
Specifications
Ice Borer
Autoclavable
500 x 25 mm
25 mm
Stainless Steel (316)
Ice, wax, and other similar materials
500 mm