missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric acid, meets analytical spec. of BP, NF, Ph. Eur., fuming, 36.5-38%, Honeywell Fluka™
Meets analytical specification of BP, NF, Ph. Eur., fuming, 36.5-38%
44.72 € - 190.52 €
Chemical Identifiers
CAS | 7647-01-0 |
---|---|
Molecular Formula | ClH |
Molecular Weight (g/mol) | 36.46 |
MDL Number | MFCD00011324 MFCD00792839 |
InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof |
PubChem CID | 313 |
ChEBI | CHEBI:17883 |
SMILES | Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15630840
|
Honeywell-Fluka
07102-1L-GL |
1 L | Glass Bottle |
44.72 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15620840
|
Honeywell-Fluka
07102-1L |
1 L | Plastic Bottle |
44.99 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15650840
|
Honeywell-Fluka
07102-2.5L-GL |
2.5 L | Glass Bottle |
66.39 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15640840
|
Honeywell-Fluka
07102-2.5L |
2.5 L | Plastic Bottle |
66.39 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15600850
|
Honeywell-Fluka
07102-6X1L-GL |
6 x 1 L | Glass Bottle | ||||||
15690840
|
Honeywell-Fluka
07102-6X1L |
6 x 1 L | Plastic Bottle | ||||||
15680840
|
Honeywell-Fluka
07102-4X2.5L-GL |
4 x 2.5 L | Glass Bottle | ||||||
15670840
|
Honeywell-Fluka
07102-4X2.5L |
4 x 2.5 L | Plastic Bottle | ||||||
15660840
|
Honeywell-Fluka
07102-30KG |
30 kg | Paper / Poly Bag | ||||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7647-01-0 | |
36.46 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
313 | |
Cl |
ClH | |
MFCD00011324 MFCD00792839 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
CHEBI:17883 |
Specifications
Yellow | |
36.5 to 38% | |
≤0.002% (as SO4) | |
ClH | |
1789 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
Cl | |
36.46 | |
CHEBI:17883 | |
Hydrochloric Acid, Meets analytical specification of Ph. Eur.BP, NF |
7647-01-0 | |
1.2g/mL at 25°C | |
3.23psi (21.1°C), 7.93psi (37.7°C) | |
MFCD00011324 MFCD00792839 | |
1098214 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
hydrogen chloride | |
313 | |
36.46g/mol |
Safety and Handling
P261-P280-P305 + P351 + P338-P310
H290-H314-H335
missing translation for 'einecsNumber' : 231-595-7
missing translation for 'rtecsNumber' : MW4025000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Hydrochloric acid
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.