missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid, meets analytical spec. of BP, NF, Ph. Eur., fuming, 36.5-38%, Honeywell Fluka™

Product Code. 15680840
Click to view available options
Quantity:
1 L
2.5 L
30 kg
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
Paper / Poly Bag
Plastic Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Meets analytical specification of BP, NF, Ph. Eur., fuming, 36.5-38%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl

Specifications

Color Yellow
Density 1.2g/mL at 25°C
Packaging Glass Bottle
Residue after Ignition ≤0.002% (as SO4)
Vapor Pressure 3.23psi (21.1°C), 7.93psi (37.7°C)
Quantity 4 x 2.5 L
UN Number UN1789
IUPAC Name hydrogen chloride
Formula Weight 36.46
Percent Purity 36.5 to 38%
Chemical Name or Material Hydrochloric Acid, Meets analytical specification of Ph. Eur.BP, NF
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hydrochloric acid
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'