missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-17 Quantikine ELISA Kit

Human IL-17 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

752.00 € - 4290.00 €

Specifications

Assay Range 31.20 - 2000pg/mL
Assay Sensitivity 15pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18772304
View Documents
R&D Systems™
D1700
1 Plate
752.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18762274
View Documents
R&D Systems™
S1700
6 Plates
4290.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Human IL-17 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human IL-17/IL-17A in cell culture supernates (100 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul), citrate plasma (100 ul).
Specifications

Specifications

31.20 - 2000pg/mL
HRP
Solid Phase Sandwich ELISA
Natural and Recombinant Human IL-17
ELISA
Cell Culture Supernates (100uL),Serum (100uL),EDTA Plasma (100uL),Heparin Plasma (100uL),Citrate Plasma (100uL)
CTLA8, CTLA-8, CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL17, IL17A, IL-17 A, IL-17 Acytotoxic T-lymphocyte-associated protein 8, IL-17 CTLA-8, IL17interleukin-17 A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17 A
15pg/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum
Human
3605
Store the unopened product at 2° to 8°C. Do not use past expiration date.
Videos
SDS
Documents

Documents

Product Certifications