missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IL-17 Quantikine ELISA Kit
Human IL-17 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand: R&D Systems™ S1700
This item is not returnable.
View return policy
Description
The Human IL-17 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human IL-17/IL-17A in cell culture supernates (100 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul), citrate plasma (100 ul).Specifications
31.20 - 2000pg/mL | |
HRP | |
Solid Phase Sandwich ELISA | |
Natural and Recombinant Human IL-17 | |
Human | |
3605 | |
Cell Culture Supernates (100uL),Serum (100uL),EDTA Plasma (100uL),Heparin Plasma (100uL),Citrate Plasma (100uL) | |
CTLA8, CTLA-8, CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL17, IL17A, IL-17 A, IL-17 Acytotoxic T-lymphocyte-associated protein 8, IL-17 CTLA-8, IL17interleukin-17 A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17 A |
15pg/mL | |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. | |
Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum | |
Sufficient materials to run ELISAs on at least six 96-well plates | |
ELISA | |
6 Plates | |
Store the unopened product at 2° to 8°C. Do not use past expiration date. |