missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIPK2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | HIPK2 |
---|---|
Concentration | 0.31 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16816875
|
Proteintech
55408-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16806875
|
Proteintech
55408-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HIPK2 is a conserved serine/threonine nuclear kinase that interacts with homeodomain transcription factors.Specifications
HIPK2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
Q9H2X6, Q9QZR5 | |
15258, 28996 | |
Peptide | |
Primary | |
-20°C | |
HIPK2 |
0.31 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
hHIPk2, HIPK2, PRO0593 | |
HIPK2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title