missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIPK2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55408-1-AP-20UL
This item is not returnable.
View return policy
Description
HIPK2 is a conserved serine/threonine nuclear kinase that interacts with homeodomain transcription factors.Specifications
HIPK2 | |
Polyclonal | |
Unconjugated | |
HIPK2 | |
hHIPk2, HIPK2, PRO0593 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15258, 28996 | |
-20°C | |
Liquid |
Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9H2X6, Q9QZR5 | |
HIPK2 | |
Peptide | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction