missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexane, Laboratory Reagent, ≥95%, Honeywell™
Laboratory Reagent,≥95%
35.12 € - 3558.10 €
Chemical Identifiers
CAS | 110-54-3 |
---|---|
Molecular Formula | C6H14 |
Molecular Weight (g/mol) | 86.18 |
MDL Number | MFCD02179311 |
InChI Key | VLKZOEOYAKHREP-UHFFFAOYSA-N |
Synonym | n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes |
PubChem CID | 8058 |
ChEBI | CHEBI:29021 |
IUPAC Name | hexane |
SMILES | CCCCCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15601870
|
Honeywell Research Chemicals
208752-500ML |
500 mL | Glass Bottle | ||||||
15641860
|
Honeywell Research Chemicals
208752-1L |
1 L | Glass Bottle |
67.79 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15651860
|
Honeywell Research Chemicals
208752-2.5L |
2.5 L | Glass Bottle |
139.82 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15611870
|
Honeywell Research Chemicals
208752-5L |
5 L | Aluminum Bottle | ||||||
15621870
|
Honeywell Research Chemicals
208752-6X1L |
6 x 1 L | Glass Bottle |
353.89 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15681860
|
Honeywell Research Chemicals
208752-4X2.5L |
4 x 2.5 L | Glass Bottle |
473.17 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15621860
|
Honeywell Research Chemicals
208752-10L |
10 L | Poly Pail | ||||||
15691860
|
Honeywell Research Chemicals
208752-4X5L |
4 x 5 L | Aluminum Bottle | ||||||
15661860
|
Honeywell Research Chemicals
208752-20L |
20 L | Stainless Steel Can | ||||||
15671860
|
Honeywell Research Chemicals
208752-25L |
25 L | Stainless Steel Can |
690.40 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15631860
|
Honeywell Research Chemicals
208752-190L |
190 L | Metal Drum | ||||||
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Chemical Identifiers
110-54-3 | |
86.18 | |
VLKZOEOYAKHREP-UHFFFAOYSA-N | |
8058 | |
hexane |
C6H14 | |
MFCD02179311 | |
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
CHEBI:29021 | |
CCCCCC |
Specifications
-95°C | |
Colorless | |
69°C | |
−26°C (−14.8°F) | |
Liquid | |
C6H14 | |
MFCD02179311 | |
UN1208 | |
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
CCCCCC | |
86.18 | |
CHEBI:29021 | |
≥95% | |
Hexane |
110-54-3 | |
0.659g/mL at 25°C | |
0.077 | |
453°F | |
n20/D 1.375(lit.) | |
CH3(CH2)4CH3 | |
132mmHg (20°C), 256mmHg (37.7°C), 5.2psi (37.7°C) | |
1730733 | |
VLKZOEOYAKHREP-UHFFFAOYSA-N | |
hexane | |
8058 | |
86.18g/mol | |
Laboratory Reagent |
Safety and Handling
P210-P260-P280-P301 + P310-P370 + P378-P403 + P235
H225-H304-H315-H336-H361f-H373-H411
missing translation for 'dotInformation' : Class 3 - PG 2 - Hexanes
missing translation for 'einecsNumber' : 203-777-6
missing translation for 'rtecsNumber' : MN9275000