missing translation for 'onlineSavingsMsg'
Learn More

Hexane, Laboratory Reagent, ≥95%, Honeywell™

Product Code. 15621860
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
10 L
20 L
25 L
4 x 2.5 L
4 x 5 L
6 x 1 L
190 L
Packaging:
Aluminum Bottle
Glass Bottle
Metal Drum
Poly Pail
Stainless Steel Can
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Laboratory Reagent,≥95%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Melting Point -95°C
Color Colorless
Density 0.659g/mL at 25°C
Boiling Point 69°C
Quantity 10 L
Explosion Limit 0.077
Flash Point −26°C (−14.8°F)
Ignition Point 453°F
Packaging Poly Pail
Physical Form Liquid
Refractive Index n20/D 1.375(lit.)
Linear Formula CH3(CH2)4CH3
Vapor Pressure 132mmHg (20°C), 256mmHg (37.7°C), 5.2psi (37.7°C)
UN Number UN1208
Beilstein 1730733
Formula Weight 86.18g/mol
Percent Purity ≥95%
Grade Laboratory Reagent
Chemical Name or Material Hexane
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'