Learn More
Hexamethylditin, 97%, Thermo Scientific Chemicals
52.90 € - 552.00 €
Chemical Identifiers
CAS | 661-69-8 |
---|---|
Molecular Formula | C6H18Sn2 |
Molecular Weight (g/mol) | 327.63 |
MDL Number | MFCD00008277 |
InChI Key | CCRMAATUKBYMPA-UHFFFAOYSA-N |
Synonym | hexamethylditin, hexamethyldistannane, distannane, hexamethyl, pennsalt td 5032, stannane, hexamethyldi, ch3 3sn 2, dimer, bis, tin, hexamethyldi, distannane, hexamethyl-8ci,9ci |
PubChem CID | 6327618 |
SMILES | C[Sn](C)(C)[Sn](C)(C)C |
Description
It is used to prepare aryl tin compounds for microwave-assisted Stille cross-coupling with halo pyridines or copper-mediated O-arylation of phenols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used to prepare aryl tin compounds for microwave-assisted Stille cross-coupling with halo pyridines or copper-mediated O-arylation of phenols.
Solubility
Not miscible or difficult to mix in water.
Notes
Air Sensitive. Keep away from oxidizing agents, heat and air. Store under inert gas.
Chemical Identifiers
661-69-8 | |
327.63 | |
CCRMAATUKBYMPA-UHFFFAOYSA-N | |
6327618 |
C6H18Sn2 | |
MFCD00008277 | |
hexamethylditin, hexamethyldistannane, distannane, hexamethyl, pennsalt td 5032, stannane, hexamethyldi, ch3 3sn 2, dimer, bis, tin, hexamethyldi, distannane, hexamethyl-8ci,9ci | |
C[Sn](C)(C)[Sn](C)(C)C |
Specifications
661-69-8 | |
1.58 | |
61°C (142°F) | |
C6H18Sn2 | |
1.541 | |
UN2788 | |
Air sensitive | |
Not miscible or difficult to mix in water. | |
C[Sn](C)(C)[Sn](C)(C)C | |
327.63 | |
327.59 | |
Hexamethylditin |
23°C | |
85°C (45 mmHg) | |
Characteristic | |
(CH3)3SnSn(CH3)3 | |
MFCD00008277 | |
3600327 | |
hexamethylditin, hexamethyldistannane, distannane, hexamethyl, pennsalt td 5032, stannane, hexamethyldi, ch3 3sn 2, dimer, bis, tin, hexamethyldi, distannane, hexamethyl-8ci,9ci | |
CCRMAATUKBYMPA-UHFFFAOYSA-N | |
trimethyltin | |
6327618 | |
97% |
Safety and Handling
GHS H Statement
H300-H310-H227
Fatal if swallowed.
Fatal in contact with skin.
Combustible liquid.
P210-P235-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P312-P314-P330-P361-P363-P370+P378q-P501c
H227-H300+H310+H330-H335-H336-H373
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
missing translation for 'einecsNumber' : 211-549-2
missing translation for 'rtecsNumber' : WH8280000
missing translation for 'tsca' : No
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.