missing translation for 'onlineSavingsMsg'
Learn More

Hexamethylditin, 97%, Thermo Scientific Chemicals

Product Code. 11458277
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used to prepare aryl tin compounds for microwave-assisted Stille cross-coupling with halo pyridines or copper-mediated O-arylation of phenols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to prepare aryl tin compounds for microwave-assisted Stille cross-coupling with halo pyridines or copper-mediated O-arylation of phenols.

Solubility
Not miscible or difficult to mix in water.

Notes
Air Sensitive. Keep away from oxidizing agents, heat and air. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 661-69-8
Molecular Formula C6H18Sn2
Molecular Weight (g/mol) 327.63
MDL Number MFCD00008277
InChI Key CCRMAATUKBYMPA-UHFFFAOYSA-N
Synonym hexamethylditin, hexamethyldistannane, distannane, hexamethyl, pennsalt td 5032, stannane, hexamethyldi, ch3 3sn 2, dimer, bis, tin, hexamethyldi, distannane, hexamethyl-8ci,9ci
PubChem CID 6327618
SMILES C[Sn](C)(C)[Sn](C)(C)C

Specifications

Melting Point 23°C
Density 1.58
Boiling Point 85°C (45 mmHg)
Flash Point 61°C (142°F)
Odor Characteristic
Linear Formula (CH3)3SnSn(CH3)3
Refractive Index 1.541
Quantity 25 g
UN Number UN2788
Beilstein 3600327
Sensitivity Air sensitive
Solubility Information Not miscible or difficult to mix in water.
IUPAC Name trimethyltin
Formula Weight 327.59
Percent Purity 97%
Chemical Name or Material Hexamethylditin
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.