missing translation for 'onlineSavingsMsg'
Learn More

HES5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00 € - 410.00 €

Specifications

Antigen HES5
Concentration 0.25 mg/mL
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16852035
View Documents
Proteintech
22666-1-AP-20UL
20 μL
145.00 €
20µL
Please to purchase this item. Need a web account? Register with us today!
16842035
View Documents
Proteintech
22666-1-AP-150UL
150 μL
410.00 €
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Hairy and Enhancer-of-split homologues 5 (HES5) isa helix-loop-helix transcriptional factor that negatively regulates cell differentiation during embryogenesis. It contains one basic helix-loop-helix (bHLH) domain and one Orange domain. HES5 is reported to be a down stream effector of the Notch Signaling Pathway and is expressed in advanced ovarian serous adenocarcinomas as well as in cervical squamous carcinoma. Reports also suggest that HES5 is abundantly expressed in osteoarthritis along with Notch1 and Jagged1.
Specifications

Specifications

HES5
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Mouse, Human, Rat
P70120, Q03062, Q5TA89
15208, 388585, 79225
HES5 Fusion Protein Ag18270
Primary
-20°C
HES5
0.25 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
bHLHb38, Hairy and enhancer of split 5, HES5, Transcription factor HES 5
HES5
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications