Learn More
HES5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22666-1-AP-150UL
 View more versions of this product
Product Code. 16842035
  • 410.00 € / 150µL
Explore more special offers
Description
Description
Hairy and Enhancer-of-split homologues 5 (HES5) isa helix-loop-helix transcriptional factor that negatively regulates cell differentiation during embryogenesis. It contains one basic helix-loop-helix (bHLH) domain and one Orange domain. HES5 is reported to be a down stream effector of the Notch Signaling Pathway and is expressed in advanced ovarian serous adenocarcinomas as well as in cervical squamous carcinoma. Reports also suggest that HES5 is abundantly expressed in osteoarthritis along with Notch1 and Jagged1.
Specifications
Specifications
HES5
Polyclonal
Unconjugated
HES5
bHLHb38, Hairy and enhancer of split 5, HES5, Transcription factor HES 5
Rabbit
Antigen Affinity Chromatography
RUO
15208, 388585, 79225
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P70120, Q03062, Q5TA89
HES5
HES5 Fusion Protein Ag18270
150 μL
Primary
Human, Mouse, Rat