Learn More
Brand™ Wide-mouth Square HDPE Bottles
Manufactured from translucent HDPE. BRAND™ Wide-mouth Square HDPE Bottles save space on tight counters, benches, and cabinets. Tamper-proof polypropylene closure includes sealing cone with press-off retaining ring. Ideal for use as a storage container.
1.74 € - 10.35 €
Specifications
Color Translucent
Includes Press-off retaining ring
Material HDPE
Closure Color Blue
Closure Included Yes
View More Specs
Products 7
Product Code Brand Capacity (Metric) Thread Size (Metric) Dimensions (L x W x H) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Thread Size (Metric) Dimensions (L x W x H) Price Quantity & Availability  
11613129
View Documents
Brand™
130412
100 mL GL 32 45 x 45 x 101mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11623129
View Documents
Brand™
130414
250 mL GL 45 60 x 60 x 127mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11633129
View Documents
Brand™
130418
500 mL GL 54 75 x 75 x 161mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11643129
View Documents
Brand™
130422
1000 mL GL 54 85 x 85 210mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11653129
View Documents
Brand™
130424
1500 mL GL 80 111 x 111 x 209mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10121871
View Documents
Brand™
130426
2500 mL GL 80 122 x 122 x 258mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11663129
View Documents
Brand™
130428
4000 mL GL 80 142 x 142 x 294mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Manufactured from translucent HDPE.
  • Tamper-proof polypropylene closure includes sealing cone with press-off retaining ring.
  • Rectangular shape saves space on tight counters, benches, and cabinets.
  • Ideal for use as a storage container.
Specifications
Specifications
Translucent
HDPE
Yes
Square
Polypropylene
Press-off retaining ring
Blue
Wide
Square Bottle
Screw Cap with Sealing Cone