Learn More
Buerkle™ Graduated SAN Cylinder with Embossed Gradations
Measuring cylinder, tall shape, according to DIN 12 680(Glass)/ISO 6706, with hexagonal base. For use in laboratories, chemists, in workshops, during outdoor field work and wherever volumetric measurements have to be made during production
28.65 € - 34.35 €
Specifications
Base Type Hexagonal
Certifications/Compliance DIN 12681, ISO 6706
Class B
Type Graduated Cylinder
Shape Round
View More Specs
Products 2
Product Code Brand Capacity (Metric) Graduations Price Quantity & Availability  
Product Code Brand Capacity (Metric) Graduations Price Quantity & Availability  
17200684
View Documents
Buerkle™
7506-0500
500 mL
28.65 €
Each
In Stock
to see stock.
Add to basket
10730834
View Documents
Buerkle™
7506-1000
1000 mL 10 mL
34.35 €
Each
In Stock
to see stock.
Add to basket
Description
Description
  • SAN, crystal clear
  • Embossed graduation with rings at the main points
  • Adjusted to “In”
Specifications
Specifications
Hexagonal
B
Round
Embossed graduation
DIN 12681, ISO 6706
Graduated Cylinder
Chemists, During Outdoor Field Work and Wherever Volumetric Measurements Have to Be Made During Production, Laboratories, Workshop