missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycogen, from Oysters, Ultrapure, Thermo Scientific Chemicals
48.87 € - 1075.00 €
Chemical Identifiers
CAS | 9005-79-2 |
---|---|
Molecular Formula | C24H42O21 |
Molecular Weight (g/mol) | 666.579 |
MDL Number | MFCD00081547 |
InChI Key | BYSGBSNPRWKUQH-UJDJLXLFSA-N |
Synonym | glycogen, 2 r,3 r,4 s,5 s,6 r-2-2 r,3 s,4 r,5 r,6 r-4,5-dihydroxy-6-2 r,3 s,4 r,5 r,6 s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxy-2-2 s,3 r,4 s,5 s,6 r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy methyl oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, glycogen from bovine liver, 2 r,3 r,4 s,5 s,6 r-2-2 r,3 s,4 r,5 r,6 r-4,5-dihydroxy-6-2 r,3 s,4 r,5 r,6 s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxy-2-2 s,3 r,4 s,5 s,6 r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, 2 s,3 r,4 s,5 s,6 r-2-2 r,3 s,4 r,5 r,6 r-4,5-dihydroxy-3-2 r,3 r,4 s,5 s,6 r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy-6-2 r,3 s,4 r,5 r,6 s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxyoxan-2-yl methoxy-6-hydroxymethyl oxane-3,4,5-triol, 4-o-4-o-alpha-d-glucopyranosyl-6-o-alpha-d-glucopyranosyl-alpha-d-glucopyranosyl-alpha-d-glucopyranose |
PubChem CID | 439177 |
ChEBI | CHEBI:28087 |
IUPAC Name | (2 S,3 R,4 S,5 S,6 R)-2-[[(2 R,3 S,4 R,5 R,6 R)-4,5-dihydroxy-3-[(2 R,3 R,4 S,5 S,6 R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-6-[(2 R,3 S,4 R,5 R,6 S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol |
SMILES | C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3C(OC(C(C3O)O)O)CO)O)O)OC4C(C(C(C(O4)CO)O)O)O)O)O)O)O |
Description
Chemical Identifiers
9005-79-2 | |
666.579 | |
BYSGBSNPRWKUQH-UJDJLXLFSA-N | |
439177 | |
(2 S,3 R,4 S,5 S,6 R)-2-[[(2 R,3 S,4 R,5 R,6 R)-4,5-dihydroxy-3-[(2 R,3 R,4 S,5 S,6 R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-6-[(2 R,3 S,4 R,5 R,6 S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol |
C24H42O21 | |
MFCD00081547 | |
glycogen, 2 r,3 r,4 s,5 s,6 r-2-2 r,3 s,4 r,5 r,6 r-4,5-dihydroxy-6-2 r,3 s,4 r,5 r,6 s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxy-2-2 s,3 r,4 s,5 s,6 r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy methyl oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, glycogen from bovine liver, 2 r,3 r,4 s,5 s,6 r-2-2 r,3 s,4 r,5 r,6 r-4,5-dihydroxy-6-2 r,3 s,4 r,5 r,6 s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxy-2-2 s,3 r,4 s,5 s,6 r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, 2 s,3 r,4 s,5 s,6 r-2-2 r,3 s,4 r,5 r,6 r-4,5-dihydroxy-3-2 r,3 r,4 s,5 s,6 r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy-6-2 r,3 s,4 r,5 r,6 s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxyoxan-2-yl methoxy-6-hydroxymethyl oxane-3,4,5-triol, 4-o-4-o-alpha-d-glucopyranosyl-6-o-alpha-d-glucopyranosyl-alpha-d-glucopyranosyl-alpha-d-glucopyranose | |
CHEBI:28087 | |
C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3C(OC(C(C3O)O)O)CO)O)O)OC4C(C(C(C(O4)CO)O)O)O)O)O)O)O |
Specifications
9005-79-2 | |
MFCD00081547 | |
BYSGBSNPRWKUQH-UJDJLXLFSA-N | |
(2 S,3 R,4 S,5 S,6 R)-2-[[(2 R,3 S,4 R,5 R,6 R)-4,5-dihydroxy-3-[(2 R,3 R,4 S,5 S,6 R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-6-[(2 R,3 S,4 R,5 R,6 S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol | |
439177 | |
Ultrapure | |
Glycogen |
C24H42O21 | |
glycogen, 2 r,3 r,4 s,5 s,6 r-2-2 r,3 s,4 r,5 r,6 r-4,5-dihydroxy-6-2 r,3 s,4 r,5 r,6 s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxy-2-2 s,3 r,4 s,5 s,6 r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy methyl oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, glycogen from bovine liver, 2 r,3 r,4 s,5 s,6 r-2-2 r,3 s,4 r,5 r,6 r-4,5-dihydroxy-6-2 r,3 s,4 r,5 r,6 s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxy-2-2 s,3 r,4 s,5 s,6 r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, 2 s,3 r,4 s,5 s,6 r-2-2 r,3 s,4 r,5 r,6 r-4,5-dihydroxy-3-2 r,3 r,4 s,5 s,6 r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy-6-2 r,3 s,4 r,5 r,6 s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxyoxan-2-yl methoxy-6-hydroxymethyl oxane-3,4,5-triol, 4-o-4-o-alpha-d-glucopyranosyl-6-o-alpha-d-glucopyranosyl-alpha-d-glucopyranosyl-alpha-d-glucopyranose | |
C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3C(OC(C(C3O)O)O)CO)O)O)OC4C(C(C(C(O4)CO)O)O)O)O)O)O)O | |
666.579 | |
CHEBI:28087 | |
Powder |
Safety and Handling
H500
missing translation for 'einecsNumber' : 232-683-8
missing translation for 'rtecsNumber' : MC2700000
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title