missing translation for 'onlineSavingsMsg'
Learn More

Glycogen, from Oysters, Ultrapure, Thermo Scientific Chemicals

Product Code. 15782808
Click to view available options
Quantity:
5 g
100 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 9005-79-2
Molecular Formula C24H42O21
Molecular Weight (g/mol) 666.579
MDL Number MFCD00081547
InChI Key BYSGBSNPRWKUQH-UJDJLXLFSA-N
Synonym glycogen, 2 r,3 r,4 s,5 s,6 r-2-2 r,3 s,4 r,5 r,6 r-4,5-dihydroxy-6-2 r,3 s,4 r,5 r,6 s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxy-2-2 s,3 r,4 s,5 s,6 r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy methyl oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, glycogen from bovine liver, 2 r,3 r,4 s,5 s,6 r-2-2 r,3 s,4 r,5 r,6 r-4,5-dihydroxy-6-2 r,3 s,4 r,5 r,6 s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxy-2-2 s,3 r,4 s,5 s,6 r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, 2 s,3 r,4 s,5 s,6 r-2-2 r,3 s,4 r,5 r,6 r-4,5-dihydroxy-3-2 r,3 r,4 s,5 s,6 r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy-6-2 r,3 s,4 r,5 r,6 s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxyoxan-2-yl methoxy-6-hydroxymethyl oxane-3,4,5-triol, 4-o-4-o-alpha-d-glucopyranosyl-6-o-alpha-d-glucopyranosyl-alpha-d-glucopyranosyl-alpha-d-glucopyranose
PubChem CID 439177
ChEBI CHEBI:28087
IUPAC Name (2 S,3 R,4 S,5 S,6 R)-2-[[(2 R,3 S,4 R,5 R,6 R)-4,5-dihydroxy-3-[(2 R,3 R,4 S,5 S,6 R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-6-[(2 R,3 S,4 R,5 R,6 S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3C(OC(C(C3O)O)O)CO)O)O)OC4C(C(C(C(O4)CO)O)O)O)O)O)O)O

Specifications

Packaging Plastic bottle
Quantity 100 g
Physical Form Powder
Chemical Name or Material Glycogen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'