Learn More
Hecht Karl™ Glass Rolled-Rim Vial
Designed with a rolled rim. Karl Hecht™ Glass Vials are ideal for storing small volumes of liquids.
Brand:  Hecht Karl™ 42783016
 View more versions of this product
Product Code. 10339451
  • 75.00 € / Pack of 154
Explore more special offers
Description
Description
  • Manufactured with glass for optimum visibility
  • Snap cap included
  • Available in various sizes to fit laboratory needs
Specifications
Specifications
Clear
Glass
154
Snap Cap
154 Pk.
24 mm
10 mL
Plastic
41 mm