missing translation for 'onlineSavingsMsg'
Learn More

Gelatin, Granular, Thermo Scientific Chemicals

Gelatin, CAS # 9000-70-8, also known as gelatin type A, derived from porcine skin, is a gelling compound used in electrophoresis and in vitro cell cultures.

47.74 € - 162.81 €

Chemical Identifiers

CAS 9000-70-8
MDL Number MFCD00081638
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10075660
SDS View Documents
Thermo Scientific Acros
410875000
500 g Plastic bottle
47.74 €
500g
Please to purchase this item. Need a web account? Register with us today!
10453655
SDS View Documents
Thermo Scientific Acros
410870025
2.5 kg Plastic bottle
162.81 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Gelatin derived from porcine skin is a powder created from the acidic digestion of collagen and is also known as gelatin type A

• It comprises heterogeneous mixture of water-soluble proteins of high average molecular masses, including glycine, proline and hydroxyproline

Applications

• Gelatin has been used in in vitro coating cell culture to improve attachment of cell lines • It is added to PCR reactions in order to stabilize DNA Taq polymerase

• It can be used as a blocking reagent in several biological techniques including western blotting, ELISA, and immunochemistry

• It can be applied in microencapsulation assays to delivery vehicle for release of active biomolecules since it is a biocompatible polymer

• It is used as a component of media for species differentiation in bacteriology and in light microscope autoradiography

• It has been used in the generation of scaffolds for tissue engineering assays

Specifications

Chemical Identifiers

9000-70-8
MFCD00081638

Specifications

9000-70-8
3.8 to 7.6 (1% soln., about 55°C) (PHEUR)
Authentic
15, 4415
Crystalline Powder
Beige to Yellow
13% max. (105°C)
MFCD00081638
Analysis
Gelatin
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 232-554-6

missing translation for 'rtecsNumber' : LX8580000

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.