missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals G3335
116.00 € - 182.00 €
Chemical Identifiers
CAS | 36099-95-3 |
---|---|
Molecular Formula | C16H19N3O5, C16H19N3O5 |
Synonym | H-Trp-Glu-OH |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15496439
|
Thermo Scientific Chemicals
J64528.MF |
50 mg | |||||||
15486439
|
Thermo Scientific Chemicals
J64528.MC |
100 mg |
182.00 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A cell-permeable dipeptide that acts as a selective and reversible peroxisome proliferator-activated receptor antagonist
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- A cell-permeable dipeptide that acts as a selective and reversible peroxisome proliferator-activated receptor antagonist
Chemical Identifiers
36099-95-3 | |
C16H19N3O5, C16H19N3O5 | |
H-Trp-Glu-OH |
Specifications
White | |
A cell-permeable dipeptide that acts as a selective and reversible peroxisome proliferator-activated receptor antagonist | |
C16H19N3O5, C16H19N3O5 | |
H-Trp-Glu-OH | |
Signal Transduction Reagent | |
PWIQCLSQVQBOQV-UHFFFAOYNA-N | |
2-[2-amino-3-(1H-indol-3-yl)propanamido]pentanedioic acid |
2°C to 8°C | |
36099-95-3 | |
333.34g/mol | |
Light Sensitive | |
Solid | |
NC(CC1=CNC2=CC=CC=C12)C(=O)NC(CCC(O)=O)C(O)=O | |
333.34 |
Safety and Handling
Recommended Storage : Keep cold
RUO – Research Use Only