missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals G3335

Product Code. 15496439
Click to view available options
Quantity:
50 mg
100 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

A cell-permeable dipeptide that acts as a selective and reversible peroxisome proliferator-activated receptor antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A cell-permeable dipeptide that acts as a selective and reversible peroxisome proliferator-activated receptor antagonist
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36099-95-3
Molecular Formula C16H19N3O5, C16H19N3O5
Synonym H-Trp-Glu-OH

Specifications

Quantity 50 mg
Description White
Content And Storage 2°C to 8°C
For Use With (Application) A cell-permeable dipeptide that acts as a selective and reversible peroxisome proliferator-activated receptor antagonist
Formula Weight 333.34g/mol
Sensitivity Light Sensitive
Product Type Signal Transduction Reagent
Form Solid
InChI Key PWIQCLSQVQBOQV-UHFFFAOYNA-N
SMILES NC(CC1=CNC2=CC=CC=C12)C(=O)NC(CCC(O)=O)C(O)=O
IUPAC Name 2-[2-amino-3-(1H-indol-3-yl)propanamido]pentanedioic acid
Molecular Weight (g/mol) 333.34
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'