missing translation for 'onlineSavingsMsg'
Learn More

Florisil™, Honeywell Fluka™

100-200 mesh

123.57 € - 381.05 €

Chemical Identifiers

CAS 1343-88-0
Molecular Formula MgO3Si
Molecular Weight (g/mol) 100.39
MDL Number MFCD00078249
InChI Key ZADYMNAVLSWLEQ-UHFFFAOYSA-N
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15652080
View Documents Promotion Details
Honeywell-Fluka
220736-250G
250 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15642080
View Documents Promotion Details
Honeywell-Fluka
220736-1KG
1 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

1343-88-0
100.39
ZADYMNAVLSWLEQ-UHFFFAOYSA-N
MgO3Si
MFCD00078249

Specifications

1343-88-0
MFCD00078249
NONH for all modes of transport
100.39
Florisil™
MgO3Si
100 to 200mesh
ZADYMNAVLSWLEQ-UHFFFAOYSA-N
100.39g/mol
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 215-681-1

SDS
Documents

Documents

Product Certifications