missing translation for 'onlineSavingsMsg'
Learn More

Florisil™, Honeywell Fluka™

Product Code. 15642080
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

100-200 mesh

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1343-88-0
Molecular Formula MgO3Si
Molecular Weight (g/mol) 100.39
MDL Number MFCD00078249
InChI Key ZADYMNAVLSWLEQ-UHFFFAOYSA-N

Specifications

Quantity 1 kg
Mesh Size 100 to 200mesh
UN Number NONH for all modes of transport
Formula Weight 100.39g/mol
Chemical Name or Material Florisil™
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'