missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Double Cytology Funnel with White Filter Cards
For the Shandon™ CytoSpin™ Centrifuge
Brand: Fisherbrand™ 10353
This item is not returnable.
View return policy
Description
- Functional equivalent of the Shandon Double Cytofunnel™
- Fully compatible with Shandon CytoSpin Centrifuge
- White filter card is pre-attached and is designed for single use only
- Funnel sealing caps included
- For sample volumes of up to 0.25mL or less
- Provides two deposition areas on each slide
Specifications
Shandon CytoSpin Centrifuge | |
White Filter Card; Sealing Caps | |
500/Cs. | |
Up to 0.25 mL |