missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-amidinoacetate hydrochloride, 95%, Thermo Scientific Chemicals

29.70 € - 462.30 €

Chemical Identifiers

CAS 57508-48-2
Molecular Formula C5H11ClN2O2
Molecular Weight (g/mol) 166.605
MDL Number MFCD06797615
InChI Key VOHFLYOSVGWQOS-UHFFFAOYSA-N
Synonym ethyl 3-amino-3-iminopropanoate hydrochloride, ethyl 2-amidinoacetate hydrochloride, 3-amino-3-iminopropanoic acid ethyl ester hydrochloride, ethyl 2-carbamimidoylacetate hydrochloride, 3-amino-3-imino-propanoic acid, ethyl ester hcl, carbamimidoyl-acetic acid ethyl ester hydrochloride, ethyl 2-amidinoacetate hcl, ethyl 3-amino-3-iminopropanate hydrochloride, ethyl 3-amino-3-iminopropanoate hcl, 3-amino-3-iminopropionic acid ethyl ester hydrochloride
PubChem CID 15555354
IUPAC Name ethyl 3-amino-3-iminopropanoate;hydrochloride
SMILES CCOC(=O)CC(=N)N.Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15401419
View Documents
Thermo Scientific Alfa Aesar
H64288.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15411419
View Documents
Thermo Scientific Alfa Aesar
H64288.06
5 g
104.52 €
5g
Please to purchase this item. Need a web account? Register with us today!
15421419
View Documents
Thermo Scientific Alfa Aesar
H64288.14
25 g
462.30 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

57508-48-2
166.605
VOHFLYOSVGWQOS-UHFFFAOYSA-N
15555354
CCOC(=O)CC(=N)N.Cl
C5H11ClN2O2
MFCD06797615
ethyl 3-amino-3-iminopropanoate hydrochloride, ethyl 2-amidinoacetate hydrochloride, 3-amino-3-iminopropanoic acid ethyl ester hydrochloride, ethyl 2-carbamimidoylacetate hydrochloride, 3-amino-3-imino-propanoic acid, ethyl ester hcl, carbamimidoyl-acetic acid ethyl ester hydrochloride, ethyl 2-amidinoacetate hcl, ethyl 3-amino-3-iminopropanate hydrochloride, ethyl 3-amino-3-iminopropanoate hcl, 3-amino-3-iminopropionic acid ethyl ester hydrochloride
ethyl 3-amino-3-iminopropanoate;hydrochloride

Specifications

57508-48-2
C5H11ClN2O2
3694030
VOHFLYOSVGWQOS-UHFFFAOYSA-N
ethyl 3-amino-3-iminopropanoate;hydrochloride
15555354
95%
Ethyl 2-amidinoacetate hydrochloride
105°C to 108°C
MFCD06797615
ethyl 3-amino-3-iminopropanoate hydrochloride, ethyl 2-amidinoacetate hydrochloride, 3-amino-3-iminopropanoic acid ethyl ester hydrochloride, ethyl 2-carbamimidoylacetate hydrochloride, 3-amino-3-imino-propanoic acid, ethyl ester hcl, carbamimidoyl-acetic acid ethyl ester hydrochloride, ethyl 2-amidinoacetate hcl, ethyl 3-amino-3-iminopropanate hydrochloride, ethyl 3-amino-3-iminopropanoate hcl, 3-amino-3-iminopropionic acid ethyl ester hydrochloride
CCOC(=O)CC(=N)N.Cl
166.605
166.6
Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.