missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-amidinoacetate hydrochloride, 95%, Thermo Scientific Chemicals

Product Code. 15401419
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57508-48-2
Molecular Formula C5H11ClN2O2
Molecular Weight (g/mol) 166.605
MDL Number MFCD06797615
InChI Key VOHFLYOSVGWQOS-UHFFFAOYSA-N
Synonym ethyl 3-amino-3-iminopropanoate hydrochloride, ethyl 2-amidinoacetate hydrochloride, 3-amino-3-iminopropanoic acid ethyl ester hydrochloride, ethyl 2-carbamimidoylacetate hydrochloride, 3-amino-3-imino-propanoic acid, ethyl ester hcl, carbamimidoyl-acetic acid ethyl ester hydrochloride, ethyl 2-amidinoacetate hcl, ethyl 3-amino-3-iminopropanate hydrochloride, ethyl 3-amino-3-iminopropanoate hcl, 3-amino-3-iminopropionic acid ethyl ester hydrochloride
PubChem CID 15555354
IUPAC Name ethyl 3-amino-3-iminopropanoate;hydrochloride
SMILES CCOC(=O)CC(=N)N.Cl

Specifications

Melting Point 105°C to 108°C
Quantity 1 g
Beilstein 3694030
Formula Weight 166.6
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Ethyl 2-amidinoacetate hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.