missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPHA2 Mouse anti-Human, Unconjugated, Clone: 1A9C3, Proteintech
Mouse Monoclonal Antibody
140.00 € - 399.00 €
Specifications
Antigen | EPHA2 |
---|---|
Clone | 1A9C3 |
Concentration | 1000 μg/mL |
Applications | Western Blot, Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16828895
|
Proteintech
66736-1-IG-20UL |
20 μL |
140.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16818895
|
Proteintech
66736-1-IG-150UL |
150 μL |
399.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The EPHA2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human EPHA2. This antibody recognizes human antigen. The EPHA2 antibody has been validated for the following applications: FC, WB, ELISA analysis.
Specifications
EPHA2 | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
1969 | |
Primary |
1A9C3 | |
Western Blot, Flow Cytometry | |
Unconjugated | |
Human | |
IgG1 |