Learn More
EPHA2 Mouse anti-Human, Unconjugated, Clone: 1A9C3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66736-1-IG-150UL
This item is not returnable.
View return policy
Description
The EPHA2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human EPHA2. This antibody recognizes human antigen. The EPHA2 antibody has been validated for the following applications: FC, WB, ELISA analysis.
Specifications
EPHA2 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Flow Cytometry | |
1A9C3 | |
Unconjugated | |
150 μL | |
1969 | |
IgG1 |