missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EED Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | EED |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16896564
|
Proteintech
16818-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16886564
|
Proteintech
16818-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The EED protein (WAIT-1 or WD protein associated with integrin cytoplasmic tails-1), also called embryonic ectoderm development, is a member of the superfamily of WD-40 repeat proteins and widely conserved polycomb group (PcG) family of proteins. The polycomb group (PcG) is a large and evolutionarily conserved set of genes whose products act in multimeric complexes to modify histones, which are then thought to cause stable and heritable states of transcriptional repression. EED shuttles between the nucleus and the plasma membrane and can interact with the cytoplasmic tail of integrin Beta-7 subunit. EED exerted an antiviral activity at the late stage of HIV-1 replication.Specifications
EED | |
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
O75530 | |
8726 | |
EED Fusion Protein Ag10358 | |
Primary | |
-20°C | |
EED |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
EED, embryonic ectoderm development, HEED, Polycomb protein EED, WAIT 1, WAIT1 | |
EED | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |