missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EED Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16818-1-AP-20UL
This item is not returnable.
View return policy
Description
The EED protein (WAIT-1 or WD protein associated with integrin cytoplasmic tails-1), also called embryonic ectoderm development, is a member of the superfamily of WD-40 repeat proteins and widely conserved polycomb group (PcG) family of proteins. The polycomb group (PcG) is a large and evolutionarily conserved set of genes whose products act in multimeric complexes to modify histones, which are then thought to cause stable and heritable states of transcriptional repression. EED shuttles between the nucleus and the plasma membrane and can interact with the cytoplasmic tail of integrin Beta-7 subunit. EED exerted an antiviral activity at the late stage of HIV-1 replication.Specifications
EED | |
Polyclonal | |
Unconjugated | |
EED | |
EED, embryonic ectoderm development, HEED, Polycomb protein EED, WAIT 1, WAIT1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8726 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75530 | |
EED | |
EED Fusion Protein Ag10358 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |