missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAH2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | DNAH2 |
---|---|
Concentration | 467 μg/mL |
Applications | Immunohistochemistry, Immunofluorescence, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16878694
|
Proteintech
18841-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16868694
|
Proteintech
18841-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The DNAH2 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human DNAH2. This antibody recognizes human antigen. The DNAH2 antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
DNAH2 | |
Immunohistochemistry, Immunofluorescence, ELISA | |
Unconjugated | |
Human | |
IgG |
467 μg/mL | |
Polyclonal | |
Rabbit | |
146754 | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title