Learn More
DNAH2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18841-1-AP-150UL
This item is not returnable.
View return policy
Description
The DNAH2 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human DNAH2. This antibody recognizes human antigen. The DNAH2 antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
DNAH2 | |
Polyclonal | |
Unconjugated | |
150 μL | |
146754 | |
IgG |
Immunohistochemistry, Immunofluorescence, ELISA | |
467 μg/mL | |
Rabbit | |
Primary | |
Human |