Learn More
Fisherbrand™ Disposable Powder-free Non-sterile Extra Protection Nitrile Examination Gloves
Resists a wide range of chemicals and resistant to puncture. Fisherbrand™ Disposable Powder-free Non-sterile Extra Protection Nitrile Examination Gloves are latex free.
28.90 € - 32.30 €
Specifications
Certifications/Compliance Conforms to EN 455 parts 1 and 2
Color Purple
Disposable Single-use
Latex Free Yes
Material Nitrile
View More Specs
Products 4
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
10769824
View Documents
Fisherbrand™
FB69294
Large This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10611783
View Documents
Fisherbrand™
FB69292
Small This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10090812
View Documents
Fisherbrand™
FB69291
X-Small This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10000822
View Documents
Fisherbrand™
FB69293
Medium This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • 100% nitrile-resistant to a wide range of chemicals and three times more puncture resistant than latex
  • Ergonomically designed with textured fingertips for improved grip
  • Latex, protein and powder free to eliminate Type I allergies
  • 0.10mm thickness
  • Non-detectable residual accelerators
  • Exceeds the requirements of EN 374 PPE Category III
  • Conforms to EN 455 parts 1 and 2
Specifications
Specifications
Conforms to EN 455 parts 1 and 2
Single-use
Nitrile
Nitrile Examination Glove
0.10 mm
50/Pk.
30 cm
Purple
Yes
Powder Free
Non-sterile
Textured
Puncture, wide range of chemicals
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Fisherbrand™ Disposable Powder-free Non-sterile Extra Protection Nitrile Examination Gloves