Learn More
Fisherbrand™ Disposable Powder-free Non-sterile Extra Protection Nitrile Examination Gloves
Product Code. 10611783
Click to view available options
Size:
X-Small
Small
Medium
Large
Resists a wide range of chemicals and resistant to puncture. Fisherbrand™ Disposable Powder-free Non-sterile Extra Protection Nitrile Examination Gloves are latex free.
  • 100% nitrile-resistant to a wide range of chemicals and three times more puncture resistant than latex
  • Ergonomically designed with textured fingertips for improved grip
  • Latex, protein and powder free to eliminate Type I allergies
  • 0.10mm thickness
  • Non-detectable residual accelerators
  • Exceeds the requirements of EN 374 PPE Category III
  • Conforms to EN 455 parts 1 and 2
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Conforms to EN 455 parts 1 and 2
Color Purple
Disposable Single-use
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type Nitrile Examination Glove
Size Small
Sterility Non-sterile
Thickness (Metric) 0.10 mm
Finger Style Textured
Quantity 50/Pk.
Resistant To Puncture, wide range of chemicals
Length (Metric) 30 cm