missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Disposable Petri Dishes

Designed for routine lab work, bacteriological research

Brand:  Kartell™ 0036300

183.51 EUR valid until 2024-12-31
Use promo code "21955" to get your promotional price.


 View more versions of this product

Product Code. 10717922

  • 198.00 € / Pack of 500
Estimated Shipment: 02-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Disposable planar, clear Petri dishes in PS
  • Designed for routine lab work, bacteriological research
  • Specifications

    Specifications

    CE marked, IVD
    Clear
    Polystyrene
    Sterile
    3
    500
    ß Beta-ray sterilization
    Single-use
    Round
    Petri Dish
    Routine lab work, bacteriological research
    90 mm
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers