missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Discharge Tubes for BRAND Dispensette S Bottletop Dispensers with Recirculation Valves

Discharge tubes with recirculation valves to fit BRAND Dispensettes made 2016 and later

Brand:  BRAND™ 708106

 View more versions of this product

Product Code. 15478033

  • 101.00 € / Each
Estimated Shipment: 17-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Discharge tubes come in two sizes: For instruments with nominal capacity 1 mL, 2 mL, 5 mL, and 10 mL; and for instruments with nominal capacity of 25 mL, 50 mL and 100 mL
  • Choose from standard and fine tip models
  • Dispensette S dispensers have red trim
  • Dispensette S Organic dispensers have yellow trim
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Discharge Tube
Red/White
Dispensing liquid
Discharge Tube, Safety Cap
135 mm
Autoclavable
ETFE, PTFE, Polypropylene
Dispensette™ S, nominal volumes 25, 50 and 100 mL
Fine
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers