Learn More
BRAND™ Discharge Tubes for BRAND Dispensette S Bottletop Dispensers with Recirculation Valves
Discharge tubes with recirculation valves to fit BRAND Dispensettes made 2016 and later
Brand:  BRAND™ 708119
 View more versions of this product
Product Code. 15418043
  • 104.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Discharge tubes come in two sizes: For instruments with nominal capacity 1 mL, 2 mL, 5 mL, and 10 mL; and for instruments with nominal capacity of 25 mL, 50 mL and 100 mL
  • Choose from standard and fine tip models
  • Dispensette S dispensers have red trim
  • Dispensette S Organic dispensers have yellow trim
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Discharge Tube
White/Yellow
Dispensing liquid
Discharge Tube, Safety Cap
135 mm
Autoclavable
ETFE, PTFE, Polypropylene
Dispensette™ S Organic, nominal volumes 25, 50 and 100 mL
Standard