Learn More
Dibromochloromethane, 97+%, Thermo Scientific Chemicals
34.47 € - 372.86 €
Chemical Identifiers
CAS | 124-48-1 |
---|---|
Molecular Formula | CHBr2Cl |
Molecular Weight (g/mol) | 208.277 |
MDL Number | MFCD00000820 |
InChI Key | GATVIKZLVQHOMN-UHFFFAOYSA-N |
Synonym | chlorodibromomethane, methane, dibromochloro, dibromo chloro methane, monochlorodibromomethane, cdbm, methane, chlorodibromo, unii-3t4ajr1h24, ccris 938, dibromo-chloro-methane, chlorobromoform |
PubChem CID | 31296 |
ChEBI | CHEBI:34627 |
IUPAC Name | dibromo(chloro)methane |
SMILES | C(Cl)(Br)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11418776
|
Thermo Scientific Alfa Aesar
A16938.06 |
5 g |
34.47 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11428776
|
Thermo Scientific Alfa Aesar
A16938.14 |
25 g |
108.54 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11438776
|
Thermo Scientific Alfa Aesar
A16938.22 |
100 g |
372.86 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It affords bromochlorocarbene under phase-transfer conditions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt affords bromochlorocarbene under phase-transfer conditions.
Solubility
Not miscible or difficult to mix in water.
Notes
Moisture & Light Sensitive. Store away from light, water/moisture, oxidizing agents and heat. Store under inert gas.
Chemical Identifiers
124-48-1 | |
208.277 | |
GATVIKZLVQHOMN-UHFFFAOYSA-N | |
31296 | |
dibromo(chloro)methane |
CHBr2Cl | |
MFCD00000820 | |
chlorodibromomethane, methane, dibromochloro, dibromo chloro methane, monochlorodibromomethane, cdbm, methane, chlorodibromo, unii-3t4ajr1h24, ccris 938, dibromo-chloro-methane, chlorobromoform | |
CHEBI:34627 | |
C(Cl)(Br)Br |
Specifications
124-48-1 | |
2.42 | |
CHBr2Cl | |
MFCD00000820 | |
Light and moisture sensitive | |
chlorodibromomethane, methane, dibromochloro, dibromo chloro methane, monochlorodibromomethane, cdbm, methane, chlorodibromo, unii-3t4ajr1h24, ccris 938, dibromo-chloro-methane, chlorobromoform | |
GATVIKZLVQHOMN-UHFFFAOYSA-N | |
dibromo(chloro)methane | |
31296 | |
208.28 | |
Dibromochloromethane |
-22°C | |
115°C to 118°C | |
1.5475 | |
1731046 | |
14,2135 | |
Not miscible or difficult to mix in water. | |
C(Cl)(Br)Br | |
208.277 | |
CHEBI:34627 | |
98% |
Safety and Handling
GHS H Statement
H341-H302
Suspected of causing genetic defects.
Harmful if swallowed.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 204-704-0
missing translation for 'rtecsNumber' : PA6360000
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold; Store under Nitrogen
- Dibromochloromethane
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Chlorodibromomethane
RUO – Research Use Only