missing translation for 'onlineSavingsMsg'
Learn More

Dibromochloromethane, 97+%, Thermo Scientific Chemicals

Product Code. 11428776
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It affords bromochlorocarbene under phase-transfer conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It affords bromochlorocarbene under phase-transfer conditions.

Solubility
Not miscible or difficult to mix in water.

Notes
Moisture & Light Sensitive. Store away from light, water/moisture, oxidizing agents and heat. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-48-1
Molecular Formula CHBr2Cl
Molecular Weight (g/mol) 208.277
MDL Number MFCD00000820
InChI Key GATVIKZLVQHOMN-UHFFFAOYSA-N
Synonym chlorodibromomethane, methane, dibromochloro, dibromo chloro methane, monochlorodibromomethane, cdbm, methane, chlorodibromo, unii-3t4ajr1h24, ccris 938, dibromo-chloro-methane, chlorobromoform
PubChem CID 31296
ChEBI CHEBI:34627
IUPAC Name dibromo(chloro)methane
SMILES C(Cl)(Br)Br

Specifications

Melting Point -22°C
Density 2.42
Boiling Point 115°C to 118°C
Refractive Index 1.5475
Quantity 25 g
Beilstein 1731046
Sensitivity Light and moisture sensitive
Merck Index 14,2135
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 208.28
Percent Purity 98%
Chemical Name or Material Dibromochloromethane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'