missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHRS3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00 € - 399.00 €
Specifications
Antigen | DHRS3 |
---|---|
Concentration | 0.25 mg/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16874544
|
Proteintech
15393-1-AP-20UL |
20 μL |
140.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16864544
|
Proteintech
15393-1-AP-150UL |
150 μL |
399.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DHRS3 catalyze the oxidation/reduction of a wide range of substrates, including retinoids and steroids.Specifications
DHRS3 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
O75911, O88876 | |
20148, 313689, 9249 | |
DHRS3 Fusion Protein Ag7589 | |
Primary | |
-20°C | |
DHRS3 |
0.25 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
DD83.1, DHRS3, RDH17, retSDR1, Rsdr1, SDR1, SDR16C1 | |
Dhrs3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |