Learn More
DHRS3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15393-1-AP-20UL
 View more versions of this product
Product Code. 16874544
  • 145.00 € / 20µL
Explore more special offers
Description
Description
DHRS3 catalyze the oxidation/reduction of a wide range of substrates, including retinoids and steroids.
Specifications
Specifications
DHRS3
Polyclonal
Unconjugated
DHRS3
DD83.1, DHRS3, RDH17, retSDR1, Rsdr1, SDR1, SDR16C1
Rabbit
Antigen Affinity Chromatography
RUO
20148, 313689, 9249
-20°C
Liquid
Western Blot, Immunoprecipitation
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O75911, O88876
Dhrs3
DHRS3 Fusion Protein Ag7589
20 μL
Primary
Rat, Mouse, Human