missing translation for 'onlineSavingsMsg'
Learn More

Desyl chloride, 98%, Thermo Scientific Chemicals

35.28 € - 308.54 €

Chemical Identifiers

CAS 447-31-4
Molecular Formula C14H11ClO
Molecular Weight (g/mol) 230.69
MDL Number MFCD00000858
InChI Key RXDYOLRABMJTEF-UHFFFAOYNA-N
Synonym desyl chloride, 2-chloro-2-phenylacetophenone, alpha-chlorodeoxybenzoin, 1,2-diphenyl-2-chloroethanone, ethanone, 2-chloro-1,2-diphenyl, alpha-chlorobenzyl phenyl ketone, 2-chloro-1,2-diphenylethone, acetophenone, alpha-chloro-alpha-phenyl, 2-chloro-1,2-diphenyl-ethanone, acetophenone, 2-chloro-2-phenyl
PubChem CID 95343
IUPAC Name 2-chloro-1,2-diphenylethanone
SMILES ClC(C(=O)C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11490407
View Documents
Thermo Scientific Alfa Aesar
B21778.06
5 g
35.28 €
5g
Please to purchase this item. Need a web account? Register with us today!
11400417
View Documents
Thermo Scientific Alfa Aesar
B21778.14
25 g
109.55 €
25g
Please to purchase this item. Need a web account? Register with us today!
11410417
View Documents
Thermo Scientific Alfa Aesar
B21778.22
100 g
308.54 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

447-31-4
230.69
RXDYOLRABMJTEF-UHFFFAOYNA-N
95343
ClC(C(=O)C1=CC=CC=C1)C1=CC=CC=C1
C14H11ClO
MFCD00000858
desyl chloride, 2-chloro-2-phenylacetophenone, alpha-chlorodeoxybenzoin, 1,2-diphenyl-2-chloroethanone, ethanone, 2-chloro-1,2-diphenyl, alpha-chlorobenzyl phenyl ketone, 2-chloro-1,2-diphenylethone, acetophenone, alpha-chloro-alpha-phenyl, 2-chloro-1,2-diphenyl-ethanone, acetophenone, 2-chloro-2-phenyl
2-chloro-1,2-diphenylethanone

Specifications

447-31-4
C14H11ClO
UN3261
desyl chloride, 2-chloro-2-phenylacetophenone, alpha-chlorodeoxybenzoin, 1,2-diphenyl-2-chloroethanone, ethanone, 2-chloro-1,2-diphenyl, alpha-chlorobenzyl phenyl ketone, 2-chloro-1,2-diphenylethone, acetophenone, alpha-chloro-alpha-phenyl, 2-chloro-1,2-diphenyl-ethanone, acetophenone, 2-chloro-2-phenyl
ClC(C(=O)C1=CC=CC=C1)C1=CC=CC=C1
230.69
230.69
Desyl chloride
64°C to 69°C
MFCD00000858
744474
RXDYOLRABMJTEF-UHFFFAOYNA-N
2-chloro-1,2-diphenylethanone
95343
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P261-P271-P280-P302+P352-P304+P340-P312-P363-P501c

H312+H332-H335

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II

missing translation for 'einecsNumber' : 207-181-7

missing translation for 'rtecsNumber' : AM6825000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.