missing translation for 'onlineSavingsMsg'
Learn More

Desyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 11400417
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 447-31-4
Molecular Formula C14H11ClO
Molecular Weight (g/mol) 230.69
MDL Number MFCD00000858
InChI Key RXDYOLRABMJTEF-UHFFFAOYNA-N
Synonym desyl chloride, 2-chloro-2-phenylacetophenone, alpha-chlorodeoxybenzoin, 1,2-diphenyl-2-chloroethanone, ethanone, 2-chloro-1,2-diphenyl, alpha-chlorobenzyl phenyl ketone, 2-chloro-1,2-diphenylethone, acetophenone, alpha-chloro-alpha-phenyl, 2-chloro-1,2-diphenyl-ethanone, acetophenone, 2-chloro-2-phenyl
PubChem CID 95343
IUPAC Name 2-chloro-1,2-diphenylethanone
SMILES ClC(C(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 64°C to 69°C
Quantity 25 g
UN Number UN3261
Beilstein 744474
Formula Weight 230.69
Percent Purity 98%
Chemical Name or Material Desyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.