Learn More
D-3-Bromocamphor-8-sulfonic acid ammonium salt, 98%, Thermo Scientific Chemicals
31.40 € - 425.00 €
Chemical Identifiers
CAS | 14575-84-9 |
---|---|
Molecular Formula | C10H18BrNO4S |
Molecular Weight (g/mol) | 328.221 |
MDL Number | MFCD00167438 |
InChI Key | GFBVBBRNPGPROZ-ZEMXJPTRSA-N |
Synonym | d-3-bromocamphor-8-sulfonic acid ammonium salt, +-3-bromocamphor-8-sulfonic acid ammonium salt, 1r-endo,anti-+-3-bromocamphor-8-sulfonic acid ammonium salt, 1r,3s,4s,7r-3-bromo-1,7-dimethyl-2-oxobicyclo 2.2.1 heptan-7-yl methanesulfonic acid amine, ammonium 1r,3s,4s,7r-3-bromo-1,7-dimethyl-2-oxobicyclo 2.2.1 heptan-7-yl methanesulfonate |
PubChem CID | 71306887 |
IUPAC Name | azane;[(1S,2S,4R,7R)-2-bromo-4,7-dimethyl-3-oxo-7-bicyclo[2.2.1]heptanyl]methanesulfonic acid |
SMILES | CC12CCC(C1(C)CS(=O)(=O)O)C(C2=O)Br.N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11335289
|
Thermo Scientific Alfa Aesar
A19886.06 |
5 g | N/A | ||||||
11345289
|
Thermo Scientific Alfa Aesar
A19886.14 |
25 g |
115.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11355289
|
Thermo Scientific Alfa Aesar
A19886.22 |
100 g | N/A | ||||||
Description
D-3-Bromocamphor-8-sulfonic acid ammonium salt is used in optical resolutions. It is also used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsD-3-Bromocamphor-8-sulfonic acid ammonium salt is used in optical resolutions. It is also used as pharmaceutical intermediates.
Solubility
Soluble in Methanol and Water (200 g/L) at 25°C.
Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store at -20°C.
Chemical Identifiers
14575-84-9 | |
328.221 | |
GFBVBBRNPGPROZ-ZEMXJPTRSA-N | |
71306887 | |
CC12CCC(C1(C)CS(=O)(=O)O)C(C2=O)Br.N |
C10H18BrNO4S | |
MFCD00167438 | |
d-3-bromocamphor-8-sulfonic acid ammonium salt, +-3-bromocamphor-8-sulfonic acid ammonium salt, 1r-endo,anti-+-3-bromocamphor-8-sulfonic acid ammonium salt, 1r,3s,4s,7r-3-bromo-1,7-dimethyl-2-oxobicyclo 2.2.1 heptan-7-yl methanesulfonic acid amine, ammonium 1r,3s,4s,7r-3-bromo-1,7-dimethyl-2-oxobicyclo 2.2.1 heptan-7-yl methanesulfonate | |
azane;[(1S,2S,4R,7R)-2-bromo-4,7-dimethyl-3-oxo-7-bicyclo[2.2.1]heptanyl]methanesulfonic acid |
Specifications
14575-84-9 | |
C10H18BrNO4S | |
d-3-bromocamphor-8-sulfonic acid ammonium salt, +-3-bromocamphor-8-sulfonic acid ammonium salt, 1r-endo,anti-+-3-bromocamphor-8-sulfonic acid ammonium salt, 1r,3s,4s,7r-3-bromo-1,7-dimethyl-2-oxobicyclo 2.2.1 heptan-7-yl methanesulfonic acid amine, ammonium 1r,3s,4s,7r-3-bromo-1,7-dimethyl-2-oxobicyclo 2.2.1 heptan-7-yl methanesulfonate | |
GFBVBBRNPGPROZ-ZEMXJPTRSA-N | |
azane;[(1S,2S,4R,7R)-2-bromo-4,7-dimethyl-3-oxo-7-bicyclo[2.2.1]heptanyl]methanesulfonic acid | |
71306887 | |
98% |
∼284°C (decomposition) | |
MFCD00167438 | |
Soluble in Methanol and Water (200g/L) at 25°C. | |
CC12CCC(C1(C)CS(=O)(=O)O)C(C2=O)Br.N | |
328.221 | |
328.23 | |
D-3-Bromocamphor-8-sulfonic acid ammonium salt |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 238-616-9
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only