missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-3-Bromocamphor-8-sulfonic acid ammonium salt, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
D-3-Bromocamphor-8-sulfonic acid ammonium salt is used in optical resolutions. It is also used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
D-3-Bromocamphor-8-sulfonic acid ammonium salt is used in optical resolutions. It is also used as pharmaceutical intermediates.
Solubility
Soluble in Methanol and Water (200 g/L) at 25°C.
Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store at -20°C.
D-3-Bromocamphor-8-sulfonic acid ammonium salt is used in optical resolutions. It is also used as pharmaceutical intermediates.
Solubility
Soluble in Methanol and Water (200 g/L) at 25°C.
Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store at -20°C.
Specifications
Specifications
Melting Point | ∼284°C (decomposition) |
Quantity | 25 g |
Solubility Information | Soluble in Methanol and Water (200g/L) at 25°C. |
Formula Weight | 328.23 |
Percent Purity | 98% |
Chemical Name or Material | D-3-Bromocamphor-8-sulfonic acid ammonium salt |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'