missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 7-Specific Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | Cytokeratin 7-Specific |
---|---|
Concentration | 180 μg/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16816624
|
Proteintech
16857-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16806624
|
Proteintech
16857-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Cytokeratin 7-specific antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Cytokeratin 7-specific. This antibody recognizes human, mouse antigen. The Cytokeratin 7-specific antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Cytokeratin 7-Specific | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Human, Mouse | |
IgG |
180 μg/mL | |
Polyclonal | |
Rabbit | |
3855 | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title